Merck
  • Facilitation of brain mitochondrial activity by 5-aminolevulinic acid in a mouse model of Alzheimer's disease.
Facilitation of brain mitochondrial activity by 5-aminolevulinic acid in a mouse model of Alzheimer's disease.
Nutritional neuroscience (2016-06-23)
Chiori Omori, Rika Motodate, Yuzuha Shiraki, Kyoko Chiba, Yuriko Sobu, Ayano Kimura, Tadashi Nakaya, Hikaru Kondo, Satoshi Kurumiya, Toru Tanaka, Kazuo Yamamoto, Motowo Nakajima, Toshiharu Suzuki, Saori Hata
ABSTRACT
The activities of mitochondrial enzymes, which are essential for neural function, decline with age and in age-related disease. In particular, the activity of cytochrome c oxidase (COX/complex IV) decreases in patients with Alzheimer's disease (AD). COX, a mitochondrial inner membrane protein complex that contains heme, plays an essential role in the electron transport chain that generates ATP. Heme synthesis begins with 5-aminolevulinic acid (5-ALA) in mitochondria. 5-ALA synthetase is the rate-limiting enzyme in heme synthesis, suggesting that supplementation with 5-ALA might help preserve mitochondrial activity in the aged brain. We administered a diet containing 5-ALA to triple-transgenic AD (3xTg-AD) model mice for 6 months, starting at 3 months of age. COX activity and protein expression, as well as mitochondrial membrane potential, were significantly higher in brains of 5-ALA-fed mice than in controls. Synaptotagmin protein levels were also significantly higher in 5-ALA-fed mice, suggesting improved preservation of synapses. Although brain Aβ levels tended to decrease in 5-ALA-fed mice, we observed no other significant changes in other biochemical and pathological hallmarks of AD. Nevertheless, our study suggests that daily oral administration of 5-ALA could preserve mitochondrial enzyme activities in the brains of aged individuals, thereby contributing to the preservation of neural activity.
Sigma-Aldrich
Cytochrome c Oxidase Assay Kit, sufficient for 100 tests, soluble and membrane bound mitochondria
Sigma-Aldrich
JC-1, powder or solid (Crystals)