- Fitness of ALS-Inhibitors Herbicide Resistant Population of Loose Silky Bentgrass (
Fitness of ALS-Inhibitors Herbicide Resistant Population of Loose Silky Bentgrass (
Herbicide resistance is an example of plant evolution caused by an increased reliance on herbicides with few sites of action to manage weed populations. This micro-evolutionary process depends on fitness, therefore the assessment of fitness differences between susceptible and resistant populations are pivotal to establish management strategies. Loose silky bentgrass (