Merck
  • Inflammasome Activation Underlying Central Nervous System Deterioration in HIV-Associated Tuberculosis.
Inflammasome Activation Underlying Central Nervous System Deterioration in HIV-Associated Tuberculosis.
The Journal of infectious diseases (2016-12-10)
Suzaan Marais, Rachel P J Lai, Katalin A Wilkinson, Graeme Meintjes, Anne O'Garra, Robert J Wilkinson
ABSTRACT
Tuberculous meningitis (TBM) is a frequent cause of meningitis in individuals with human immunodeficiency virus (HIV) infection, resulting in death in approximately 40% of affected patients. A severe complication of antiretroviral therapy (ART) in these patients is neurological tuberculosis-immune reconstitution inflammatory syndrome (IRIS), but its underlying cause remains poorly understood. To investigate the pathogenesis of TBM-IRIS, we performed longitudinal whole-blood microarray analysis of HIV-infected patients with TBM and reflected the findings at the protein level. Patients in whom TBM-IRIS eventually developed had significantly more abundant neutrophil-associated transcripts, from before development of TBM-IRIS through IRIS symptom onset. After ART initiation, a significantly higher abundance of transcripts associated with canonical and noncanonical inflammasomes was detected in patients with TBM-IRIS than in non-IRIS controls. Whole-blood transcriptome findings complement protein measurement from the site of disease, which together suggest a dominant role for the innate immune system in the pathogenesis of TBM-IRIS.
Sigma-Aldrich
2-Methyl-2-propanethiol, 99%