Merck
Electro-chemiluminescent biosensing.
Analytical and bioanalytical chemistry (2007-10-03)
Christophe A Marquette, Loïc J Blum
ABSTRACT
The present review draws a general picture of the bioanalytical applications of electro-chemiluminescent reactions (ECL). Only the two main ECL reactions-i.e. the luminol-based and Ru(bpy)(3)(2+)-based reactions-are considered for application in the fields of enzyme biosensors, immunochemical biosensors, DNA biosensors, and biochips. The mechanism, principle, and experimental conditions of these two reactions are described. Then, for each category of analytical tools, experimental set-ups and performances are presented and discussed.
Sigma-Aldrich
Cholesterol Oxidase microbial, recombinant, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Glucose dehydrogenase from Pseudomonas sp., powder, white, ≥200 U/mg
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa