Merck
Cryo-EM structures of the ATP-bound Vps4
Nature communications (2017-07-18)
Shan Sun, Lin Li, Fan Yang, Xiaojing Wang, Fenghui Fan, Mengyi Yang, Chunlai Chen, Xueming Li, Hong-Wei Wang, Sen-Fang Sui
ABSTRACT
The cellular ESCRT-III (endosomal sorting complex required for transport-III) and Vps4 (vacuolar protein sorting 4) comprise a common machinery that mediates a variety of membrane remodelling events. Vps4 is essential for the machinery function by using the energy from ATP hydrolysis to disassemble the ESCRT-III polymer into individual proteins. Here, we report the structures of the ATP-bound Vps4
Sigma-Aldrich
4-Nitrobenzyl alcohol, 99%
Sigma-Aldrich
Cyclooctatetraene, 98%
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Supelco
Malachite Green chloride, analytical standard