Merck
  • T follicular helper and T follicular regulatory cells have different TCR specificity.
T follicular helper and T follicular regulatory cells have different TCR specificity.
Nature communications (2017-04-22)
Ana Raquel Maceiras, Silvia Cristina Paiva Almeida, Encarnita Mariotti-Ferrandiz, Wahiba Chaara, Fadi Jebbawi, Adrien Six, Shohei Hori, David Klatzmann, Jose Faro, Luis Graca
ABSTRACT
Immunization leads to the formation of germinal centres (GCs) that contain both T follicular helper (Tfh) and T follicular regulatory (Tfr) cells. Whether T-cell receptor (TCR) specificity defines the differential functions of Tfh and Tfr cells is unclear. Here we show that antigen-specific T cells after immunization are preferentially recruited to the GC to become Tfh cells, but not Tfr cells. Tfh cells, but not Tfr cells, also proliferate efficiently on restimulation with the same immunizing antigen in vitro. Ex vivo TCR repertoire analysis shows that immunization induces oligoclonal expansion of Tfh cells. By contrast, the Tfr pool has a TCR repertoire that more closely resembles that of regulatory T (Treg) cells. Our data thus indicate that the GC Tfh and Tfr pools are generated from distinct TCR repertoires, with Tfh cells expressing antigen-responsive TCRs to promote antibody responses, and Tfr cells expressing potentially autoreactive TCRs to suppress autoimmunity.
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Freund′s Adjuvant, Incomplete, liquid
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥90% (PAGE), lyophilized powder