Merck
A voltage-dependent K
The Journal of cell biology (2017-05-05)
Wuyang Wang, Xiaoli Zhang, Qiong Gao, Maria Lawas, Lu Yu, Xiping Cheng, Mingxue Gu, Nirakar Sahoo, Xinran Li, Ping Li, Stephen Ireland, Andrea Meredith, Haoxing Xu
ABSTRACT
The resting membrane potential (Δψ) of the cell is negative on the cytosolic side and determined primarily by the plasma membrane's selective permeability to K
Sigma-Aldrich
Iberiotoxin, recombinant from Mesobuthus tamulus, ≥98% (HPLC), recombinant, expressed in E. coli
Sigma-Aldrich
Clofilium tosylate, >97%, solid