Merck
MICU1 drives glycolysis and chemoresistance in ovarian cancer.
Nature communications (2017-05-23)
Prabir K Chakraborty, Soumyajit Banerjee Mustafi, Xunhao Xiong, Shailendra Kumar Dhar Dwivedi, Vasyl Nesin, Sounik Saha, Min Zhang, Danny Dhanasekaran, Muralidharan Jayaraman, Robert Mannel, Kathleen Moore, Scott McMeekin, Da Yang, Rosemary Zuna, Kai Ding, Leonidas Tsiokas, Resham Bhattacharya, Priyabrata Mukherjee
ABSTRACT
Cancer cells actively promote aerobic glycolysis to sustain their metabolic requirements through mechanisms not always clear. Here, we demonstrate that the gatekeeper of mitochondrial Ca
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
Sigma-Aldrich
Pyr6, ≥98% (HPLC)
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture