Merck
  • Microfluidization of Graphite and Formulation of Graphene-Based Conductive Inks.
Microfluidization of Graphite and Formulation of Graphene-Based Conductive Inks.
ACS nano (2017-01-20)
Panagiotis G Karagiannidis, Stephen A Hodge, Lucia Lombardi, Flavia Tomarchio, Nicolas Decorde, Silvia Milana, Ilya Goykhman, Yang Su, Steven V Mesite, Duncan N Johnstone, Rowan K Leary, Paul A Midgley, Nicola M Pugno, Felice Torrisi, Andrea C Ferrari
ABSTRACT
We report the exfoliation of graphite in aqueous solutions under high shear rate [∼ 10
Sigma-Aldrich
Graphene ink in water, inkjet printable
Sigma-Aldrich
Graphene ink in water, screen printable
Sigma-Aldrich
Graphene ink in water, flexo/gravure/screen printable