Merck
  • The effect of the site of laser zona opening on the complete hatching of mouse blastocysts and their cell numbers.
The effect of the site of laser zona opening on the complete hatching of mouse blastocysts and their cell numbers.
Clinical and experimental reproductive medicine (2016-10-01)
Usanee Sanmee, Waraporn Piromlertamorn, Teraporn Vutyavanich
ABSTRACT
We studied the effect of the site of laser zona opening on the complete hatching of mouse blastocysts and the cell numbers of the completely hatched blastocysts. Mouse blastocysts were randomly allocated to the inner cell mass (ICM) group (zona opening performed at the site of the ICM, n=125), the trophectoderm (TE) group (zona opening performed opposite to the ICM, n=125) and the control group (no zona opening, n=125). The rate of complete hatching of the blastocysts was not significantly different in the ICM and the TE group (84.8% vs 80.8%, respectively; p=0.402), but was significantly lower in the control group (51.2%, p<0.001). The cell numbers in the completely hatched blastocysts were comparable in the control group, the ICM group, and the TE group (69Ā±19.3, 74Ā±15.7, and 71Ā±16.8, respectively; p=0.680). These findings indicate that the site of laser zona opening did not influence the rate of complete hatching of mouse blastocysts or their cell numbers.
Sigma-Aldrich
Anti-Mouse Serum antibody produced in rabbit, whole antiserum