Merck
  • K-252a promotes survival and choline acetyltransferase activity in striatal and basal forebrain neuronal cultures.
K-252a promotes survival and choline acetyltransferase activity in striatal and basal forebrain neuronal cultures.
Journal of neurochemistry (1995-04-01)
M A Glicksman, M E Forbes, J E Prantner, N T Neff
ABSTRACT
The organic molecule K-252a promoted cell survival, neurite outgrowth, and increased choline acetyltransferase (ChAT) activity in rat embryonic striatal and basal forebrain cultures in a concentration-dependent manner. A two- to threefold increase in survival was observed at 75 nM K-252a in both systems. A single application of K-252a at culture initiation prevented substantial (> 60%) cell death that otherwise occurred after 4 days in striatal or basal forebrain cultures. A 5-h exposure of striatal or basal forebrain cells to K-252a, followed by its removal, resulted in survival equivalent to that observed in cultures continually maintained in its presence. This is in contrast to results found with a 5-h exposure of basal forebrain cultures to nerve growth factor (NGF). Acute exposure of basal forebrain cultures to K-252a, but not to NGF, increased ChAT activity, indicating that NGF was required the entire culture period for maximum activity. Striatal cholinergic and GABAergic neurons were among the neurons rescued by K-252a. Of the protein growth factors tested in striatal cultures (ciliary neurotrophic factor, neurotrophin-3, NGF, brain-derived neurotrophic factor, interleukin-2, basic fibroblast growth factor), only brain-derived neurotrophic factor promoted survival. The enhancement of survival and ChAT activity of basal forebrain and striatal neurons by K-252a defines additional populations of neurons in which survival and/or differentiation is regulated by a K-252a-responsive mechanism. The above results expand the potential therapeutic targets for these molecules for the treatment of neuro-degenerative diseases.
Sigma-Aldrich
K-252a, Ready Made Solution, from Nonomuraea longicatena, >98%
Pricing and availability is not currently available. Contact Technical Service