Merck
  • Arabidopsis florigen FT binds to diurnally oscillating phospholipids that accelerate flowering.
Arabidopsis florigen FT binds to diurnally oscillating phospholipids that accelerate flowering.
Nature communications (2014-04-05)
Yuki Nakamura, Fernando Andrés, Kazue Kanehara, Yu-chi Liu, Peter Dörmann, George Coupland
ABSTRACT
Arabidopsis FT protein is a component of florigen, which transmits photoperiodic flowering signals from leaf companion cells to the shoot apex. Here, we show that FT specifically binds phosphatidylcholine (PC) in vitro. A transgenic approach to increase PC levels in vivo in the shoot meristem accelerates flowering whereas reduced PC levels delay flowering, demonstrating that PC levels are correlated with flowering time. The early flowering is related to FT activity, because expression of FT-effector genes is increased in these plants. Simultaneous increase of FT and PC in the shoot apical meristem further stimulates flowering, whereas a loss of FT function leads to an attenuation of the effect of increased PC. Specific molecular species of PC oscillate diurnally, and night-dominant species are not the preferred ligands of FT. Elevating night-dominant species during the day delays flowering. We suggest that FT binds to diurnally changing molecular species of PC to promote flowering.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid
Pricing and availability is not currently available.
Avanti
Egg PC, Avanti Research - A Croda Brand
Pricing and availability is not currently available.
Avanti
Egg PC, Avanti Research - A Croda Brand
Pricing and availability is not currently available.
Avanti
Soy PS, Avanti Research - A Croda Brand 870336C
Pricing and availability is not currently available.
Avanti
Soy PS, Avanti Research - A Croda Brand
Pricing and availability is not currently available.