Merck
Molecular pathways: interleukin-15 signaling in health and in cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-04-17)
Anjali Mishra, Laura Sullivan, Michael A Caligiuri
ABSTRACT
Interleukin-15 (IL-15) is a proinflammatory cytokine involved in the development, survival, proliferation, and activation of multiple lymphocyte lineages utilizing a variety of signaling pathways. IL-15 utilizes three distinct receptor chains in at least two different combinations to signal and exert its effects on the immune system. The binding of IL-15 to its receptor complex activates an "immune-enhancing" signaling cascade in natural killer cells and subsets of T cells, as well as the induction of a number of proto-oncogenes. Additional studies have explored the role of IL-15 in the development and progression of cancer, notably leukemia of large granular lymphocytes, cutaneous T-cell lymphoma, and multiple myeloma. This review provides an overview of the molecular events in the IL-15 signaling pathway and the aberrancies in its regulation that are associated with chronic inflammation and cancer. We briefly explore the potential therapeutic opportunities that have arisen as a result of these studies to further the treatment of cancer. These involve both targeting the disruption of IL-15 signaling as well as IL-15-mediated enhancement of innate and antigen-specific immunity.
Sigma-Aldrich
IL-15 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Pricing and availability is not currently available.