Merck
  • Method for the simultaneous determination of free/protein malondialdehyde and lipid/protein hydroperoxides.
Method for the simultaneous determination of free/protein malondialdehyde and lipid/protein hydroperoxides.
Free radical biology & medicine (2012-10-09)
Konstantinos Grintzalis, Dimitrios Zisimopoulos, Tilman Grune, Daniela Weber, Christos D Georgiou
ABSTRACT
A simple and sensitive method is presented for the simultaneous quantification (spectrophotometric and spectrofluorimetric) of the main lipid and protein peroxidation products after their initial fractionation: free malondialdehyde (FrMDA), protein-bound malondialdehyde (PrMDA), total hydroperoxides (LOOH), and protein hydroperoxides (PrOOH). FrMDA and PrMDA (released from proteins by alkaline hydrolysis) are measured after the reaction of MDA with thiobarbituric acid (TBA) under acidic conditions, by the specific fluorimetric quantification of the resulting MDA-(TBA)2 adduct chromophore. The measurement of LOOH and PrOOH is based on the reaction of Fe(3+) (resulting from the reaction of LOOH and PrOOH with Fe(2+)) with xylenol orange (XO) and the photometric quantification of the resulting XO-Fe complex. The sensitivity of the assays for FrMDA/PrMDA and LOOH/PrOOH is 20 and 100pmol, respectively. The method was applied successfully on human plasma and can be used for the evaluation of oxidative stress in both basic and clinical research.
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, ACS reagent, 99%
Pricing and availability is not currently available.