Merck
  • A Novel Styryldehydropyridocolinium Homodimer: Synthesis and Fluorescence Properties Upon Interaction with DNA.
A Novel Styryldehydropyridocolinium Homodimer: Synthesis and Fluorescence Properties Upon Interaction with DNA.
Journal of fluorescence (2015-09-19)
Huirong Yao, Lifang Chang, Chang Liu, Xiaojie Jiao, Song He, Haijun Liu, Xianshun Zeng
ABSTRACT
A novel homodimer of the styryldehydropyridocolinium dye (TPTP) has been synthesized and characterized. Free TPTP exhibited low fluorescence quantum yield and large Stokes shift (over 160 nm) in water. However, it showed a significant fluorescence turn-on effect upon intercalation into DNA base pairs. Meanwhile, the fluorescence intensity of the intercalated structures formed by TPTP and DNA decreased quickly upon addition of deoxyribonuclease I, indicating that the dye can be used to monitor deoxyribonuclease I activity and DNA hydrolysis. Electrophoresis analysis revealed that the dye had intercalative binding to DNA and can potentially be used for DNA staining in electrophoresis. Thus, the innate nature of large Stokes shift and excellent fluorescence turn on effect upon interaction with DNA endue the dye with a wide range of applications.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from calf thymus, Type I, fibers
Sigma-Aldrich
4-(Diethylamino)salicylaldehyde, 98%