Merck
  • Light-driven generation of hydrogen: New chromophore dyads for increased activity based on Bodipy dye and Pt(diimine)(dithiolate) complexes.
Light-driven generation of hydrogen: New chromophore dyads for increased activity based on Bodipy dye and Pt(diimine)(dithiolate) complexes.
Proceedings of the National Academy of Sciences of the United States of America (2015-06-28)
Bo Zheng, Randy P Sabatini, Wen-Fu Fu, Min-Sik Eum, William W Brennessel, Lidong Wang, David W McCamant, Richard Eisenberg
ABSTRACT
New dyads consisting of a strongly absorbing Bodipy (dipyrromethene-BF2) dye and a platinum diimine dithiolate (PtN2S2) charge transfer (CT) chromophore have been synthesized and studied in the context of the light-driven generation of H2 from aqueous protons. In these dyads, the Bodipy dye is bonded directly to the benzenedithiolate ligand of the PtN2S2 CT chromophore. Each of the new dyads contains either a bipyridine (bpy) or phenanthroline (phen) diimine with an attached functional group that is used for binding directly to TiO2 nanoparticles, allowing rapid electron photoinjection into the semiconductor. The absorption spectra and cyclic voltammograms of the dyads show that the spectroscopic and electrochemical properties of the dyads are the sum of the individual chromophores (Bodipy and the PtN2S2 moieties), indicating little electronic coupling between them. Connection to TiO2 nanoparticles is carried out by sonication leading to in situ attachment to TiO2 without prior hydrolysis of the ester linking groups to acids. For H2 generation studies, the TiO2 particles are platinized (Pt-TiO2) so that the light absorber (the dyad), the electron conduit (TiO2), and the catalyst (attached colloidal Pt) are fully integrated. It is found that upon 530 nm irradiation in a H2O solution (pH 4) with ascorbic acid as an electron donor, the dyad linked to Pt-TiO2 via a phosphonate or carboxylate attachment shows excellent light-driven H2 production with substantial longevity, in which one particular dyad [4(bpyP)] exhibits the highest activity, generating ∼ 40,000 turnover numbers of H2 over 12 d (with respect to dye).
Sigma-Aldrich
2,2′-Bipyridine-4,4′-dicarboxylic acid, 98%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis