Merck
  • PvdP is a tyrosinase that drives maturation of the pyoverdine chromophore in Pseudomonas aeruginosa.
PvdP is a tyrosinase that drives maturation of the pyoverdine chromophore in Pseudomonas aeruginosa.
Journal of bacteriology (2014-05-13)
Pol Nadal-Jimenez, Gudrun Koch, Carlos R Reis, Remco Muntendam, Hans Raj, C Margot Jeronimus-Stratingh, Robbert H Cool, Wim J Quax
ABSTRACT
The iron binding siderophore pyoverdine constitutes a major adaptive factor contributing to both virulence and survival in fluorescent pseudomonads. For decades, pyoverdine production has allowed the identification and classification of fluorescent and nonfluorescent pseudomonads. Here, we demonstrate that PvdP, a periplasmic enzyme of previously unknown function, is a tyrosinase required for the maturation of the pyoverdine chromophore in Pseudomonas aeruginosa. PvdP converts the nonfluorescent ferribactin, containing two iron binding groups, into a fluorescent pyoverdine, forming a strong hexadentate complex with ferrous iron, by three consecutive oxidation steps. PvdP represents the first characterized member of a small family of tyrosinases present in fluorescent pseudomonads that are required for siderophore maturation and are capable of acting on large peptidic substrates.
Product Description
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
Levodopa, European Pharmacopoeia (EP) Reference Standard
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)