Merck
  • Incorporation of eicosatrienoic acid exerts mild anti-inflammatory properties in murine RAW264.7 cells.
Incorporation of eicosatrienoic acid exerts mild anti-inflammatory properties in murine RAW264.7 cells.
Inflammation (2014-07-06)
Szu-Jung Chen, Lu-Te Chuang, Sung-Nien Chen
ABSTRACT
Eicosatrienoic acid (Δ11,14,17-20:3; ETrA) is a rare naturally occurring n-3 polyunsaturated fatty acid (PUFA). Using murine RAW264.7 cells, the objectives were to determine how ETrA modulated phospholipid fatty acid compositions and the production of pro-inflammatory mediators. Incubation cells with ETrA dose-dependently increased the proportions of phospholipid ETrA and its metabolites to 33 % of the fatty acid total. Incorporation of ETrA also reduced the proportions of total n-6 PUFA and monounsaturated fatty acids (MUFA) by 30 and 60 %, respectively. ETrA suppressed LPS-stimulated nuclear factor-kappa B (NF-κB)-mediated nitric oxide (NO) production and inducible nitric oxide synthase (iNOS) expression. However, no such suppressive effect on the production of prostaglandin E2 (PGE2), cytokines, or expression of cyclooxygenase-2 (COX-2) was observed. As compared with ETrA, eicosapentaenoic acid (EPA) exerted a more potent anti-inflammatory effect. In conclusion, although ETrA suppresses significant NO synthesis and iNOS expression, this n-3 PUFA was a less potent anti-inflammatory agent than EPA.
Product Description
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
USP
Sulfanilamide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sulfanilamide, United States Pharmacopeia (USP) Reference Standard
Sulfanilamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
4,5-Dimethylthiazole, ≥97%, FG
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Supelco
Sodium nitrite, analytical standard
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%