Merck
  • Kinetic evaluation of glucose 1-phosphate analogues with a thymidylyltransferase using a continuous coupled enzyme assay.
Kinetic evaluation of glucose 1-phosphate analogues with a thymidylyltransferase using a continuous coupled enzyme assay.
Organic & biomolecular chemistry (2014-11-20)
S M Forget, A Jee, D A Smithen, R Jagdhane, S Anjum, S A Beaton, D R J Palmer, R T Syvitski, D L Jakeman
ABSTRACT
Cps2L, a thymidylytransferase, is the first enzyme in Streptococcus pneumoniae L-rhamnose biosynthesis and an antibacterial target. We herein report the evaluation of six sugar phosphate analogues selected to further probe Cps2L substrate tolerance. A modified continuous spectrophotometric assay was employed for facile detection of pyrophosphate (PPi) released from nucleotidylyltransfase-catalysed condensation of sugar 1-phosphates and nucleoside triphosphates to produce sugar nucleotides. Additionally, experiments using waterLOGSY NMR spectroscopy were investigated as a complimentary method to evaluate binding affinity to Cps2L.
Sigma-Aldrich
Isopentenyl pyrophosphate triammonium salt solution, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Xanthine, ≥99%
Sigma-Aldrich
Xanthine, BioUltra, ≥99%