Merck
PI3Kγ integrates cAMP and Akt signalling of the μ-opioid receptor.
British journal of pharmacology (2014-03-25)
Sreedhar Madishetti, Nadine Schneble, Christian König, Emilio Hirsch, Stefan Schulz, Jörg P Müller, Reinhard Wetzker
ABSTRACT
The μ-opioid receptor has been characterized as the main mediator of opioid signalling in neuronal cells. Opioid-induced pain suppression was originally proposed to be mediated by μ-opioid receptor-induced inhibitory effects on cAMP, which is known to mediate inflammatory hypernociception. Recent investigations revealed PI3Kγ and Akt (PKB) as additional elements of μ-opioid receptor signalling. Hence, we investigated the interaction between pronociceptive cAMP and antinociceptive PI3K/Akt signalling pathways. The human neuroblastoma cell line SK-N-LO and primary dorsal root ganglia (DRG) cells from mice were used to elucidate mediators of μ-opioid receptor signalling. In both cellular systems cAMP was manipulated by stimulation of adenylate cyclase and consequent effects on PI3K/Akt signalling were analysed. Morphine stimulated Akt phosphorylation on Ser(473) and Thr(308) in a dose- and time-dependent manner indicating a functional μ-opioid receptor/Akt signalling pathway in μ-SK-N-LO cells. This effect of morphine was suppressed by the μ-opioid receptor inhibitor, naloxone, Pertussis toxin, an inhibitor of Gi heterotrimeric G-proteins, and the pan PI3K inhibitor wortmannin. cAMP-elevating agents also suppressed μ-opioid receptor-dependent stimulation of PI3Kγ lipid kinase and Akt activities in SK-N-LO cells and DRG. The data unveil a hitherto unknown interaction of pronociceptive cAMP and antinociceptive PI3K/Akt signalling pathways in neuronal cells. PI3Kγ was identified as a mediator of the inhibitory action of cAMP on Akt in SK-N-LO cells and DRG. The data indicate that PI3Kγ has a critical role in cAMP-mediated inflammatory hypernociception and analgesic signalling via μ-opioid receptors and PI3K/Akt in neuronal cells.
Product Description
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
USP
Naloxone, United States Pharmacopeia (USP) Reference Standard
Supelco
Forskolin, analytical standard