Merck
  • Phosphorylated STAT3 and PD-1 regulate IL-17 production and IL-23 receptor expression in Mycobacterium tuberculosis infection.
Phosphorylated STAT3 and PD-1 regulate IL-17 production and IL-23 receptor expression in Mycobacterium tuberculosis infection.
European journal of immunology (2014-03-20)
Anuradha Bandaru, Kamakshi P Devalraju, Padmaja Paidipally, Rohan Dhiman, Sambasivan Venkatasubramanian, Peter F Barnes, Ramakrishna Vankayalapati, Vijayalakshmi Valluri
ABSTRACT
We studied the factors that regulate IL-23 receptor expression and IL-17 production in human tuberculosis infection. Mycobacterium tuberculosis (M. tb)-stimulated CD4(+) T cells from tuberculosis patients secreted less IL-17 than did CD4(+) T cells from healthy tuberculin reactors (PPD(+) ). M. tb-cultured monocytes from tuberculosis patients and PPD(+) donors expressed equal amounts of IL-23p19 mRNA and protein, suggesting that reduced IL-23 production is not responsible for decreased IL-17 production by tuberculosis patients. Freshly isolated and M. tb-stimulated CD4(+) T cells from tuberculosis patients had reduced IL-23 receptor and phosphorylated STAT3 (pSTAT3) expression, compared with cells from PPD(+) donors. STAT3 siRNA reduced IL-23 receptor expression and IL-17 production by CD4(+) T cells from PPD(+) donors. Tuberculosis patients had increased numbers of PD-1(+) T cells compared with healthy PPD(+) individuals. Anti-PD-1 antibody enhanced pSTAT3 and IL-23R expression and IL-17 production by M. tb-cultured CD4(+) T cells of tuberculosis patients. Anti-tuberculosis therapy decreased PD-1 expression, increased IL-17 and IFN-γ production and pSTAT3 and IL-23R expression. These findings demonstrate that increased PD-1 expression and decreased pSTAT3 expression reduce IL-23 receptor expression and IL-17 production by CD4(+) T cells of tuberculosis patients.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stat3
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Human IL-17 ELISA Kit, for serum, plasma, cell culture supernatant and urine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mouse IL-17 (IL-17A) ELISA Kit, for serum, plasma and cell culture supernatant