Merck
  • Azilsartan increases levels of IL-10, down-regulates MMP-2, MMP-9, RANKL/RANK, Cathepsin K and up-regulates OPG in an experimental periodontitis model.
Azilsartan increases levels of IL-10, down-regulates MMP-2, MMP-9, RANKL/RANK, Cathepsin K and up-regulates OPG in an experimental periodontitis model.
PloS one (2014-05-14)
Aurigena Antunes de Araújo, Hugo Varela, Gerly Anne de Castro Brito, Caroline Addison Carvalho Xavier de Medeiros, Lorena de Souza Araújo, José Heriberto Oliveira do Nascimento, Raimundo Fernandes de Araújo Júnior
ABSTRACT
The aim of this study was to evaluate the effects of azilsartan (AZT) on bone loss, inflammation, and the expression of matrix metallo proteinases (MMPs), receptor activator of nuclear factor κB ligand (RANKL), receptor activator of nuclear factor κB (RANK), osteoprotegerin (OPG), cyclooxygenase-2 (COX-2), and cathepsin K in periodontal tissue in a rat model of ligature-induced periodontitis. Male Wistar albino rats were randomly divided into 5 groups of 10 rats each: (1) nonligated, water; (2) ligated, water; (3) ligated, 1 mg/kg AZT; (4) ligated, 5 mg/kg AZT; and (5) ligated, 10 mg/kg AZT. All groups were treated with saline or AZT for 10 days. Periodontal tissues were analyzed by histopathology and immunohistochemical detection of MMP-2, MMP-9, COX-2, RANKL, RANK, OPG, and cathepsin K. Levels of IL-1β, IL-10, TNF-α, myeloperoxidase (MPO), and glutathione (GSH) were determined by ELISA. Treatment with 5 mg/kg AZT resulted in reduced MPO (p<0.05) and IL-1β (p<0.05), increased levels of IL-10 (p<0.05), and reduced expression of MMP-2, MMP-9, COX-2, RANK, RANKL, cathepsin K, and increased expression of OPG. These findings reveal that AZT increases anti-inflammatory cytokines and GSH and decreases bone loss in ligature-induced periodontitis in rats.
Sigma-Aldrich
Bovine TNFα / Tumor Necrosis Factor alpha ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Interleukin-10 from mouse, >97% (SDS-PAGE), recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for cell and tissue lysates