Merck
  • Tetrabutylammonium bisulfate: a new effective catalyst for the hydrolysis of aziridines or epoxides.
Tetrabutylammonium bisulfate: a new effective catalyst for the hydrolysis of aziridines or epoxides.
Organic & biomolecular chemistry (2003-08-21)
Ren-Hua Fan, Xue-Long Hou
ABSTRACT
Bu4NHSO4 (TBAHS) is an effective catalyst for the hydrolysis of aziridines and epoxides under mild and non-metal conditions to give the corresponding beta-amino alcohols and 1,2-diols in high yields. The catalyst can be recycled.
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)