Merck
  • Cytotoxic and anti-inflammatory prenylated benzoylphloroglucinols and xanthones from the twigs of Garcinia esculenta.
Cytotoxic and anti-inflammatory prenylated benzoylphloroglucinols and xanthones from the twigs of Garcinia esculenta.
Journal of natural products (2014-06-25)
Hong Zhang, Dan-Dan Zhang, Yuan-Zhi Lao, Wen-Wei Fu, Shuang Liang, Qing-Hong Yuan, Ling Yang, Hong-Xi Xu
ABSTRACT
Five new prenylated benzoylphloroglucinol derivatives, garciesculentones A-E (1-5), a new xanthone, garciesculenxanthone A (6), and 15 known compounds were isolated from the petroleum ether extract and the EtOAc-soluble fraction of a 80% (v/v) EtOH extract of Garcinia esculenta. The structures of the new compounds were elucidated by 1D- and 2D-NMR spectroscopic analysis and mass spectrometry. Experimental and calculated ECD and a convenient modified Mosher's method were used to determine the absolute configurations. The cytotoxicity of these compounds were evaluated by MTT assay against three human cancer cell lines (HepG2, MCF-7, and MDA-MB-231) and against normal hepatic cells (HL-7702). In addition, these isolates were evaluated for their inhibitory effects on interferon-γ plus lipopolysaccharide-induced nitric oxide production in RAW264.7 cells.
Sigma-Aldrich
Phloroglucinol, Used to detect the presence of wood fiber.
Sigma-Aldrich
Phloroglucinol, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Phloroglucinol (anhydrous), European Pharmacopoeia (EP) Reference Standard