Merck
  • Impact of prothrombin time-International Normalized Ratio on outcome of patients with septic shock receiving polymyxin B cartridge hemoperfusion.
Impact of prothrombin time-International Normalized Ratio on outcome of patients with septic shock receiving polymyxin B cartridge hemoperfusion.
Surgery (2014-06-16)
Mitsuru Ishizuka, Kazuma Tago, Keiichi Kubota
ABSTRACT
Although most patients with septic shock have a poor outcome, some may survive after blood purification treatment such as polymyxin B cartridge hemoperfusion (PMX). To explore the most significant characteristic associated with 28-day mortality in patients with septic shock receiving PMX. Between April 2006 and March 2008, 116 patients with septic shock who had received PMX in a prospectively collected multicenter collaborative study were enrolled. Uni- and multivariate analyses using the Cox proportional hazard model were performed to assess the most significant clinical characteristic that was associated with 28-day mortality. Among 33 clinicolaboratory characteristics, receiver operating characteristic (ROC) curve analyses selected 12 characteristics with recommended cutoff values such as HCO(3)(-) (≤19.8/>19.8; mEq/L), base excess (≤-5.35/>-5.35; mEq/L), diastolic blood pressure (≤48/>48 mmHg), mean arterial pressure (≤73/>73 mmHg), pH (≤7.29/>7.29), interleukin-6 (≤19,150/>19,150 pg/dL), prothrombin time-International Normalized Ratio (PT-INR; ≤2.05/>2.05), predictive value of Acute Physiology and Chronic Health Evaluation II (APACHE II; ≤0.4/>0.4), pyruvate (≤1.82/>1.82 mg/dL), APACHE II score (≤21/>21), acetate/pyruvate ratio (≤19/>19), and acetate (≤44.8/>44.8 mg/dL) on the basis of large area under the ROC curves for 28-day mortality. The results of uni- and multivariate analyses using these selected characteristics revealed that only PT-INR (≤2.05/>2.05; hazard ratio, 2.823; 95% CI, 1.243-6.412; P = .013) was associated with 28-day mortality. Survival curve analysis demonstrated a significant difference in 28-day mortality between patients with lower (≤2.05) and higher (>2.05) PT-INR (P < .001). Prolonged PT-INR is an independent risk factor for 28-day mortality in patients receiving PMX for septic shock.
Sigma-Aldrich
Polymyxin B sulfate salt
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Supelco
Polymyxin B solution, 1 mg/mL in H2O, analytical standard
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O
Polymyxin B sulfate for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications
Polymyxin B sulfate, European Pharmacopoeia (EP) Reference Standard