Merck
  • Experimental and numerical validation of the effective medium theory for the B-term band broadening in 1st and 2nd generation monolithic silica columns.
Experimental and numerical validation of the effective medium theory for the B-term band broadening in 1st and 2nd generation monolithic silica columns.
Journal of chromatography. A (2014-06-10)
Sander Deridder, Alison Vanmessen, Kazuki Nakanishi, Gert Desmet, Deirdre Cabooter
ABSTRACT
Effective medium theory (EMT) expressions for the B-term band broadening in monolithic silica columns are presented at the whole-column as well as at the mesoporous skeleton level. Given the bi-continuous nature of the monolithic medium, regular as well as inverse formulations of the EMT-expressions have been established. The established expressions were validated by applying them to a set of experimental effective diffusion (Deff)-data obtained via peak parking on a number of 1st and 2nd generation monolithic silica columns, as well as to a set of numerical diffusion simulations in a simplified monolithic column representation (tetrahedral skeleton model) with different external porosities and internal diffusion coefficients. The numerically simulated diffusion data can be very closely represented over a very broad range of zone retention factors (up to k″=80) using the established EMT-expressions, especially when using the inverse variant. The expressions also allow representing the experimentally measured effective diffusion data very closely. The measured Deff/Dmol-values were found to decrease significantly with increasing retention factor, in general going from about Deff/Dmol=0.55 to 0.65 at low k″ (k″≅1.5-3.8) to Deff/Dmol=0.25 at very high k″ (k″≅40-80). These values are significantly larger than observed in fully-porous and core-shell particles. The intra-skeleton diffusion coefficient (Dpz) was typically found to be of the order of Dpz/Dmol=0.4, compared to Dpz/Dmol=0.2-0.35 observed in most particle-based columns. These higher Dpz/Dmol values are the cause of the higher Deff/Dmol values observed. In addition, it also appears that the higher internal diffusion is linked to the higher porosity of the mesoporous skeleton that has a relatively open structure with relatively wide pores. The observed (weak) relation between Dpz/Dmol and the zone retention factor appears to be in good agreement with that predicted when applying the regular variant of the EMT-expression directly to the mesoporous skeleton level.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O