Merck
Pentachlorophenol.
Mutation research (1991-01-01)
J P Seiler
ABSTRACT
Pentachlorophenol (PCP) is a substance whose widespread use, mainly in wood protection and pulp and paper mills, has led to a substantial environmental contamination. This in turn accounts for a significant exposure of the general human population, with rather high exposure levels being attained in occupational settings. Investigations on the genotoxic activity of PCP have given rise to divergent results which would seem to make an evaluation difficult. By grouping them into 3 categories a somewhat clearer picture, allowing finally an (admittedly tentative) assessment, can be obtained. PCP does seem to be at most a weak inducer of DNA damage: it produces neither DNA-strand breaks nor clear differential toxicity to bacteria in rec-assays in the absence of metabolic activation. Also in SCE induction no increase can be observed in vivo, while PCP is found marginally active in a single in vitro experiment. Metabolic activation, however, leads to prophage induction and to DNA strand breaks in human lymphocytes, presumably through the formation of oxygen radicals. A possible further exception in this area might be the positive results in the yeast recombination tests, although their inadequate reporting makes a full evaluation difficult. PCP does not seem to induce gene (point) mutations, as most bacterial assays, the Drosophila sex-linked recessive lethal test and in vitro assays with mammalian cells did not demonstrate any effects. Marginally positive results were obtained in the mammalian spot test in vivo and in one bacterial test; the positive result in the yeast assay for cycloheximide resistance is fraught somewhat with its questionable genetic basis. PCP does, however, induce chromosomal aberrations in mammalian cells in vitro and in lymphocytes of exposed persons in vivo. Those in vivo results that were unable to provide evidence of chromosomal damage are hampered either by methodological inadequacies or by too low exposure levels. The (rodent) metabolite tetrachlorohydroquinone might be a real genotoxic agent, capable of binding to DNA and producing DNA strand breaks; this activity is probably due to semiquinone radical formation and partly mediated through active oxygen species. Since this compound has not been tested in the common bacterial and mammalian mutagenicity assays, the few ancillary results on this substance cannot be used in a meaningful human risk assessment of PCP. Furthermore, this metabolite has only been produced by human liver microsomes in vitro, but has not been detected in exposed humans in vivo.(ABSTRACT TRUNCATED AT 400 WORDS)
Sigma-Aldrich
Pentachlorophenol, 97%
Supelco
Pentachlorophenol, analytical standard
Supelco
Pentachlorophenol solution, certified reference material, 5000 μg/mL in methanol