Merck
Pentoxifylline: a drug with wide spectrum applications in dermatology.
Dermatology online journal (2008-12-20)
Omid Zargari
ABSTRACT
Pentoxifylline (PTX) is a methylxanthine derivative with a variety of anti-inflammatory effects. Currently, PTX is approved by the Food and Drug Administration for the treatment of intermittent claudication, but studies have shown that it has a variety of physiological effects at the cellular level, which may be important in treating a diverse group of diseases.
Sigma-Aldrich
Pentoxifylline, solid
Pentoxifylline, European Pharmacopoeia (EP) Reference Standard
USP
Pentoxifylline, United States Pharmacopeia (USP) Reference Standard