Merck
  • Centrin 2 is required for mouse olfactory ciliary trafficking and development of ependymal cilia planar polarity.
Centrin 2 is required for mouse olfactory ciliary trafficking and development of ependymal cilia planar polarity.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-05-03)
Guoxin Ying, Prachee Avasthi, Mavis Irwin, Cecilia D Gerstner, Jeanne M Frederick, Mary T Lucero, Wolfgang Baehr
ABSTRACT
Centrins are ancient calmodulin-related Ca(2+)-binding proteins associated with basal bodies. In lower eukaryotes, Centrin2 (CETN2) is required for basal body replication and positioning, although its function in mammals is undefined. We generated a germline CETN2 knock-out (KO) mouse presenting with syndromic ciliopathy including dysosmia and hydrocephalus. Absence of CETN2 leads to olfactory cilia loss, impaired ciliary trafficking of olfactory signaling proteins, adenylate cyclase III (ACIII), and cyclic nucleotide-gated (CNG) channel, as well as disrupted basal body apical migration in postnatal olfactory sensory neurons (OSNs). In mutant OSNs, cilia base-anchoring of intraflagellar transport components IFT88, the kinesin-II subunit KIF3A, and cytoplasmic dynein 2 appeared compromised. Although the densities of mutant ependymal and respiratory cilia were largely normal, the planar polarity of mutant ependymal cilia was disrupted, resulting in uncoordinated flow of CSF. Transgenic expression of GFP-CETN2 rescued the Cetn2-deficiency phenotype. These results indicate that mammalian basal body replication and ciliogenesis occur independently of CETN2; however, mouse CETN2 regulates protein trafficking of olfactory cilia and participates in specifying planar polarity of ependymal cilia.
Sigma-Aldrich
Amyl acetate, ≥99%, FG
Sigma-Aldrich
Pentyl acetate, 99%
Sigma-Aldrich
Pentyl acetate, puriss. p.a., ≥98.5% (GC)
Supelco
Pentyl acetate, analytical standard