Merck
  • Crystal structure of glycoside hydrolase family 127 β-l-arabinofuranosidase from Bifidobacterium longum.
Crystal structure of glycoside hydrolase family 127 β-l-arabinofuranosidase from Bifidobacterium longum.
Biochemical and biophysical research communications (2014-04-01)
Tasuku Ito, Kyo Saikawa, Seonah Kim, Kiyotaka Fujita, Akihiro Ishiwata, Sophon Kaeothip, Takatoshi Arakawa, Takayoshi Wakagi, Gregg T Beckham, Yukishige Ito, Shinya Fushinobu
ABSTRACT
Enzymes acting on β-linked arabinofuranosides have been unknown until recently, in spite of wide distribution of β-l-arabinofuranosyl oligosaccharides in plant cells. Recently, a β-l-arabinofuranosidase from the glycoside hydrolase family 127 (HypBA1) was discovered in the newly characterized degradation system of hydroxyproline-linked β-l-arabinooligosaccharides in the bacterium Bifidobacterium longum. Here, we report the crystal structure of HypBA1 in the ligand-free and β-l-arabinofuranose complex forms. The structure of HypBA1 consists of a catalytic barrel domain and two additional β-sandwich domains, with one β-sandwich domain involved in the formation of a dimer. Interestingly, there is an unprecedented metal-binding motif with Zn(2+) coordinated by glutamate and three cysteines in the active site. The glutamate residue is located far from the anomeric carbon of the β-l-arabinofuranose ligand, but one cysteine residue is appropriately located for nucleophilic attack for glycosidic bond cleavage. The residues around the active site are highly conserved among GH127 members. Based on biochemical experiments and quantum mechanical calculations, a possible reaction mechanism involving cysteine as the nucleophile is proposed.
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
DL-Arabinose, ≥98% (GC)
Sigma-Aldrich
L-Glutamic acid hydrochloride
Zinc, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, 50x50mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 2.0mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 50x50mm, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 15mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 25x25mm, thickness 6mm, as rolled, 99.99+%