Merck
  • Immunomodulatory activity of zinc peroxide (ZnO₂) and titanium dioxide (TiO₂) nanoparticles and their effects on DNA and protein integrity.
Immunomodulatory activity of zinc peroxide (ZnO₂) and titanium dioxide (TiO₂) nanoparticles and their effects on DNA and protein integrity.
Toxicology letters (2014-03-19)
Stanely Makumire, Venkata S K Chakravadhanula, Gabriele Köllisch, Engelbert Redel, Addmore Shonhai
ABSTRACT
Nanoparticles that are made from zinc and titanium oxide have found widespread applications, including their use in sunscreens. However, there is little information regarding their effects on immune cells. In the current study, we synthesized charge stabilized and "ligand free" colloid stable ZnO₂ and TiO₂ nanoparticles. Most previous published studies commonly used ZnO and TiO₂ nanoparticles. In the current study we investigated the comparative toxicity of ZnO₂ and TiO₂ nanoparticles. Therefore, our results based on ZnO₂ which is more oxidative than ZnO provides novel data on the possible toxicity of this species of nanoparticles. First, we investigated the immunomodulatory action of these nanoparticles on human peripheral blood mononuclear cells and their effects on DNA and protein integrity. A minimum concentration of ZnO₂ nanoparticles of 1 μg/mL inhibited the production of two inflammatory cytokines: interleukin-1-β and interleukin 6 by peripheral blood mononuclear cells in the presence of lipopolysaccharides. On the other hand, TiO₂ nanoparticles at a concentration range of 0.1-100 μg/mL did not present apparent toxicity to the peripheral blood mononuclear cells. ZnO₂ nanoparticles at a minimum concentration of 2 μg/mL caused DNA damage in vitro. TiO₂ nanoparticles at a concentration range of 25-100 μg/mL only caused marginal DNA damage. ZnO₂ nanoparticles at a minimum concentration of 5 μg/mL were capable of promoting aggregation of malate dehydrogenase, and facilitated its degradation at higher concentrations. Exposure of malate dehydrogenase to TiO₂ at a concentration range of 2.5-15 μg/mL did not alter the solubility of malate dehydrogenase. Altogether, our findings suggest that charge stabilized ZnO₂ nanoparticles are nascent and interact with DNA and protein and may be harmful to immune cells. In addition, the propensity of ZnO₂ nanoparticles to promote protein aggregation could facilitate the production of protein complexes that may interfere with normal immune functions.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, ≥600 units/mg protein (biuret), ammonium sulfate suspension
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, ≥400 units/mg protein (biuret), ammonium sulfate suspension
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Zinc, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, 50x50mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 2.0mm, as rolled, 99.95+%