Merck
  • Cycline efficacy on the propagation of human prions in primary cultured neurons is strain-specific.
Cycline efficacy on the propagation of human prions in primary cultured neurons is strain-specific.
The Journal of infectious diseases (2013-11-23)
Samia Hannaoui, Alexianne Gougerot, Nicolas Privat, Etienne Levavasseur, Nicolas Bizat, Jean-Jacques Hauw, Jean-Philippe Brandel, Stéphane Haïk
ABSTRACT
In prion diseases, a major issue in therapeutic research is the variability of the effect between strains. Stimulated by the report of an antiprion effect in a scrapie model and by ongoing international clinical trials using doxycycline, we studied the efficacy of cyclines against the propagation of human prions. First, we successfully propagated various Creutzfeldt-Jakob disease (CJD) isolates (sporadic, variant, and iatrogenic CJD) in neuronal cultures expressing the human prion protein. Then, we found that doxycycline was the most effective compound, with important variations between isolates. Isolates from sporadic CJD, the most common form of prion disease, showed the highest sensitivity.
Product Description
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Doxycycline hyclate, VETRANAL®, analytical standard
Sigma-Aldrich
Doxycycline hyclate
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
USP
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Doxycycline monohydrate