Merck
  • Surgery for acquired cardiovascular disease: antiseptic treatment of contaminated vein grafts.
Surgery for acquired cardiovascular disease: antiseptic treatment of contaminated vein grafts.
The Journal of cardiovascular surgery (2013-10-18)
F P Schmidt, A A Peivandi, W Kohnen, B Jansen
ABSTRACT
Saphenous vein grafts harvested for use as bypass conduits can be contaminated intraoperatively, e.g. by being inadvertently dropped to the floor of the operating room (OR). This study was performed to investigate microorganisms most likely contaminating vein grafts and to assess the possible efficacy of measures to treat potentially contaminated vein grafts antiseptically for further use. In a first step we determined the microbiological flora of the OR using surface cultures and cultures from intentionally dropped vein grafts. Several antiseptic agents (PVP-iodine 10%, octenidinhydrochloride 0.1%, polyhexanide 1%) were evaluated for their in vitro efficacy to disinfect artificially contaminated vein segments. The most promising antiseptic regimen was tested on veins contaminated in a real OR setting. Finally, we tested for possible alterations in mechanical properties of the veins caused by antiseptic treatment. Coagulase-negative staphylococci where the predominant bacteria recovered from the OR with 59.9%. Antiseptic treatment with a combination of octenidine and PVP-iodine resulted in a higher rate of negative cultures than any single agent. Treatment of 50 saphenous vein grafts contaminated in the OR with the combination regimen resulted in only 3 positive cultural results within 7 days. Mechanical tear-stress testing comparing antiseptically treated vein grafts with controls showed no difference in their resistance to tear stress. Antiseptic treatment of contaminated vein grafts was shown to be effective in a high percentage of cases without altering mechanical properties of grafts and may be an option for the surgeon in case of a contamination.
Product Description
Povidone (iodinated), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Poly(vinylpyrrolidone)–Iodine complex