Merck
  • [Simultaneous determination of trihexyphenidyl, chlorpromazine and clozapine in blood by GC-MS].
[Simultaneous determination of trihexyphenidyl, chlorpromazine and clozapine in blood by GC-MS].
Fa yi xue za zhi (2011-09-15)
Shui-Qing Zheng, Wei Wang, Chen Liang, Rong Wang, Fei-Jun Gong, Zhong-Ping Wu, Yong-Sheng Chen, Yu-Rong Zhang, Run-Sheng Zhang
ABSTRACT
To develop a method to measure trihexyphenidyl, chlorpromazine and clozapine in human blood with GC-MS. The specimens were alkalized (pH > 10) and extracted with V (benzene):V(ethyl acetate) = 1:1, and qualitatively analyzed using GC-MS-Full Scan with internal standard SKF525A. The specimens were alkalized (pH > 10) and extracted with V(benzene):V(ethyl acetate) = 1:1, and quantitatively analyzed using GC-MS-SIM with internal standard diazepam-d5. The lowest detection limits of trihexyphenidyl, chlorpromazine and clozapine were 0.3, 0.3 and 0.7 ng/mL (S/N > or = 3) respectively. The calibration curve in 20-10 000 ng/mL showed a good linear distribution. The recovery rate was 79.9% to 85.5%. The RSDs of intraday and interday were less than 5.1%. The established method was simple, sensitive and accurate for simultaneous determination of trihexyphenidyl, chlorpromazine and clozapine in human blood, and can be applied in forensic toxicological cases.
Product Description
Trihexyphenidyl hydrochloride, European Pharmacopoeia (EP) Reference Standard