Merck
  • Aliphatic beta-nitro alcohols for non-enzymatic collagen cross-linking of scleral tissue.
Aliphatic beta-nitro alcohols for non-enzymatic collagen cross-linking of scleral tissue.
Experimental eye research (2008-07-12)
David C Paik, Quan Wen, Suzanna Airiani, Richard E Braunstein, Stephen L Trokel
ABSTRACT
The success of riboflavin photochemical cross-linking of the cornea in treating keratoconus and post-surgical keratectasia has prompted interest in cross-linking scleral tissue with a potential application to stabilize myopic progression. Applying an UVA light source to the sclera is difficult, particularly in the posterior region. An alternate pharmacologic approach to scleral cross-linking may be possible. The present study was undertaken in order to identify nitrite related compounds capable of inducing scleral tissue cross-linking and to gain information regarding the possible chemical mechanisms involved. 8x4 mm strips of porcine and human sclera were incubated in various concentrations of nitrite related agents (1-100mM) at 37 degrees C. pH 7.4 was used for all experiments except those involving NaNO(2). Following a 24-96 h incubation period, the samples were tested for cross-linking effects using thermal shrinkage temperature (T(s)) analysis. Several compounds were studied including NaNO(2), 2-nitroethanol, 2-nitro-1-propanol, 3-nitro-2-pentanol, 2-nitrophenol, 2-nitroethane, 2-aminoethanol, isopentyl nitrite, DPTA/NO, DETA/NO, and urea, a nitrous acid trap. The results indicate that short chain aliphatic beta-nitro alcohols (2-nitroethanol, 2-nitro-1-propanol, and 3-nitro-2-pentanol) are particularly effective cross-linking agents at pH 7.4, showing both time and concentration dependent effects. Furthermore, nitrosation does not appear to induce tissue cross-linking. In conclusion, aliphatic beta-nitro alcohols can cross-link scleral tissue at physiologic pH and temperature. Since beta-nitro alcohols are known to have reasonable toxicity profiles, these agents could find utility as pharmacologic cross-linking agents for scleral thinning disease.
Sigma-Aldrich
2-Nitroethanol, 97%
Pricing and availability is not currently available.