Merck
Identification of S-RNase and peroxidase in petunia nectar.
Journal of plant physiology (2010-11-26)
Melissa S Hillwig, Charles Kanobe, Robert W Thornburg, Gustavo C Macintosh
ABSTRACT
Previous SDS PAGE gel analysis of the floral nectars from petunia and tobacco plants revealed significant differences in the protein patterns. Petunia floral nectar was shown to contain a number of RNase activities by in gel RNase activity assay. To identify these proteins in more detail, the bands with RNase activity were excised from gel and subjected to trypsin digestion followed by LC-MS/MS analysis. This analysis revealed that S-RNases accumulate in nectar from Petunia hybrida, where they should carry out a biological function different from self-pollen rejection. In addition, other proteins were identified by the LC-MS/MS analysis. These proteins include a peroxidase, an endochitinase, and a putative fructokinase. Each of these proteins contained a secretory signal sequence that marked them as potential nectar proteins. We developed RT-PCR assays for each of these five proteins and demonstrated that each of these proteins was expressed in the petunia floral nectary. A discussion of the role of these proteins in antimicrobial activity in nectar is presented.
Sigma-Aldrich
Chitinase from Streptomyces griseus, lyophilized powder (essentially salt free), ≥200 units/g solid