Merck
  • Small-molecular inhibitors of Ca²⁺-induced mitochondrial permeability transition (MPT) derived from muscle relaxant dantrolene.
Small-molecular inhibitors of Ca²⁺-induced mitochondrial permeability transition (MPT) derived from muscle relaxant dantrolene.
Bioorganic & medicinal chemistry (2012-10-03)
Shinpei Murasawa, Katsuya Iuchi, Shinichi Sato, Tomomi Noguchi-Yachide, Mikiko Sodeoka, Tsutomu Yokomatsu, Kosuke Dodo, Yuichi Hashimoto, Hiroshi Aoyama
ABSTRACT
A structure consisting of substituted hydantoin linked to a 5-(halophenyl)furan-2-yl group via an amide bond was identified as a promising scaffold for development of low-molecular-weight therapeutic agents to treat vascular dysfunction, including ischemia/reperfusion injury. Among the compounds synthesized, 5-(3,5-dichlorophenyl)-N-{2,4-dioxo-3-[(pyridin-3-yl)methyl]imidazolidin-1-yl}-2-furamide (17) possessed the most potent inhibitory activity against Ca(2+)-induced mitochondrial swelling. The structural development, synthesis and structure-activity relationship of these compounds are described.
Product Description
Supelco
Cyclosporin D solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.