Merck
  • Vaccenic acid-mediated reduction in cytokine production is independent of c9,t11-CLA in human peripheral blood mononuclear cells.
Vaccenic acid-mediated reduction in cytokine production is independent of c9,t11-CLA in human peripheral blood mononuclear cells.
Biochimica et biophysica acta (2012-07-04)
Anke Jaudszus, Gerhard Jahreis, Wiebke Schlörmann, Janine Fischer, Ronny Kramer, Christian Degen, Carsten Rohrer, Alexander Roth, Holger Gabriel, Dagmar Barz, Michael Gruen
ABSTRACT
The ruminant trans fatty acid vaccenic acid (tVA) favorably alters markers of inflammation. However, it is not yet clear whether these effects are attributed to its endogenous partial conversion to c9,t11-CLA, which is known to possess anti-inflammatory properties. We compared the cytokine reducing potential of tVA to c9,t11-CLA in human T-helper (Th) cells as a main source of cytokine production during inflammation. Secondly, we assessed whether a bioconversion of tVA to c9,t11-CLA via stearoyl-CoA desaturase (SCD) encoded activity takes place in peripheral blood mononuclear cells (PBMC) in order to relate the outcomes of intracellular cytokine measurement to the degree of conversion. TVA reduced the percentage of both IL-2 and TNF-α expressing Th cells significantly, but to a lesser extent compared to c9,t11-CLA, as determined by flow cytometry after alloreactive stimulation of PBMC. Pre-treatment with the selective PPARγ antagonist T0070907 largely re-established the IL-2 and TNF-α positive Th cell population in both tVA and c9,t11-CLA treated cultures. Interestingly, while the portion of tVA dose-dependently increased within the cellular lipid fraction, the initially marginal amount of c9,t11-CLA remained unaltered. However, SCD mRNA although abundantly expressed in PBMC was not regulated by tVA. Conclusively, these results suggest that the cytokine reducing effect of tVA in human T cells is independent of c9,t11-CLA, since no bioconversion occurred. Moreover, the data provide evidence that tVA mechanistically acts in a manner similar to c9,t11-CLA.
Sigma-Aldrich
trans-Vaccenic acid, ≥99% (capillary GC)