Merck
  • Supramolecular photocatalysis: insights into cucurbit[8]uril catalyzed photodimerization of 6-methylcoumarin.
Supramolecular photocatalysis: insights into cucurbit[8]uril catalyzed photodimerization of 6-methylcoumarin.
Chemical communications (Cambridge, England) (2011-05-05)
Barry C Pemberton, Raushan K Singh, Alexander C Johnson, Steffen Jockusch, José P Da Silva, Angel Ugrinov, Nicholas J Turro, D K Srivastava, J Sivaguru
ABSTRACT
Guest induced shape change of the cucurbit[8]uril cavity is likely rate limiting in the supramolecular photocatalytic cycle for CB8 mediated photodimerization of 6-methylcoumarin.
Sigma-Aldrich
6-Methylcoumarin, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
5 g
Please contact Customer Service for Availability
€42.00
25 g
Please contact Customer Service for Availability
€189.00
100 g
Please contact Customer Service for Availability
€439.00
To order products, please contact your local dealer.
Sigma-Aldrich
6-Methylcoumarin, ≥99%, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
5 g
Please contact Customer Service for Availability
€42.00
25 g
Please contact Customer Service for Availability
€189.00
100 g
Please contact Customer Service for Availability
€439.00
To order products, please contact your local dealer.