Merck
  • NMR spectral assignment of 2α- and 3β-methylhopanes and evidence for boat conformation in D ring of 17α(H),21α(H)-hopanes.
NMR spectral assignment of 2α- and 3β-methylhopanes and evidence for boat conformation in D ring of 17α(H),21α(H)-hopanes.
Magnetic resonance in chemistry : MRC (2010-10-01)
Geir Kildahl-Andersen, Hans Peter Nytoft, Jon Eigill Johansen
ABSTRACT
The full (1)H and (13)C NMR chemical shift assignment of 2α-methyl-17α(H),21β(H)-hopane is presented. This compound is formed in mature sediments from biogenic sources of 2β-methyl-17β(H),21β(H)-hopanoids, which include several cyanobacteria. In addition, full (1)H and (13)C NMR chemical shift data of all four 17,21 isomers of 3β-methylhopane have been assigned. The thermodynamically most stable 3β-configuration corresponds to that found in bacterial sources. The data presented here suggest minor corrections to the (13)C chemical assignments reported earlier for 17α(H)-hopanes. Moreover, spectral evidence indicates an unexpected ring-D boat conformation of 17α(H),21α(H)-hopanes, which may serve to explain the steric strain reported for this isomer.
Product Description
Supelco
17α(H),21β(H)-Hopane solution, 0.1 mg/mL in isooctane, analytical standard
Supelco
17β(H),21β(H)-Hopane solution, 0.1 mg/mL in isooctane, analytical standard