- Seasonal variation of biomass and bioactive alkaloid content of goldenseal, Hydrastis canadensis.
Seasonal variation of biomass and bioactive alkaloid content of goldenseal, Hydrastis canadensis.
Fitoterapia (2010-06-17)
James A Douglas, John M Follett, Graeme A Parmenter, Catherine E Sansom, Nigel B Perry, Ray A Littler
PMID20550958
ABSTRACT
Seasonal variations in biomass and alkaloid contents of goldenseal (Hydrastis canadensis) were investigated. Five-year-old plants gave 5x the yield of roots and rhizomes of two-year-old plants, and summer growth gave significant increases in root biomass but not rhizomes. Berberine contents of roots plus rhizomes did not vary significantly and were >3.4% in all samples. Hydrastine contents of 5 y roots plus rhizomes showed significant seasonal variation. These variations were due to significant changes in the hydrastine contents of the roots (1.3-1.9%), but not the rhizomes (2.2-2.8%). Goldenseal leaves plus stems had lower contents of hydrastine (0.4-0.8%) and berberine (1.0-1.5%).
Product Description
Hydrastine hydrochloride, European Pharmacopoeia (EP) Reference Standard