Merck
  • Dietary intake of plant sterols stably increases plant sterol levels in the murine brain.
Dietary intake of plant sterols stably increases plant sterol levels in the murine brain.
Journal of lipid research (2012-01-27)
Tim Vanmierlo, Oliver Weingärtner, Susanne van der Pol, Constanze Husche, Anja Kerksiek, Silvia Friedrichs, Eric Sijbrands, Harry Steinbusch, Marcus Grimm, Tobias Hartmann, Ulrich Laufs, Michael Böhm, Helga E de Vries, Monique Mulder, Dieter Lütjohann
ABSTRACT
Plant sterols such as sitosterol and campesterol are frequently administered as cholesterol-lowering supplements in food. Recently, it has been shown in mice that, in contrast to the structurally related cholesterol, circulating plant sterols can enter the brain. We questioned whether the accumulation of plant sterols in murine brain is reversible. After being fed a plant sterol ester-enriched diet for 6 weeks, C57BL/6NCrl mice displayed significantly increased concentrations of plant sterols in serum, liver, and brain by 2- to 3-fold. Blocking intestinal sterol uptake for the next 6 months while feeding the mice with a plant stanol ester-enriched diet resulted in strongly decreased plant sterol levels in serum and liver, without affecting brain plant sterol levels. Relative to plasma concentrations, brain levels of campesterol were higher than sitosterol, suggesting that campesterol traverses the blood-brain barrier more efficiently. In vitro experiments with brain endothelial cell cultures showed that campesterol crossed the blood-brain barrier more efficiently than sitosterol. We conclude that, over a 6-month period, plant sterol accumulation in murine brain is virtually irreversible.
Product Description
Avanti
campesterol-d6, campesterol-d6, powder
Sigma-Aldrich
Campesterol, ~65%