Merck
  • Cysteine proteases and cell differentiation: excystment of the ciliated protist Sterkiella histriomuscorum.
Cysteine proteases and cell differentiation: excystment of the ciliated protist Sterkiella histriomuscorum.
Eukaryotic cell (2003-12-11)
Eduardo Villalobo, Clara Moch, Ghislaine Fryd-Versavel, Anne Fleury-Aubusson, Loïc Morin
ABSTRACT
The process of excystment of Sterkiella histriomuscorum (Ciliophora, Oxytrichidae) leads in a few hours, through a massive influx of water and the resorption of the cyst wall, from an undifferentiated resting cyst to a highly differentiated and dividing vegetative cell. While studying the nature of the genes involved in this process, we isolated three different cysteine proteases genes, namely, a cathepsin B gene, a cathepsin L-like gene, and a calpain-like gene. Excystation was selectively inhibited at a precise differentiating stage by cysteine proteases inhibitors, suggesting that these proteins are specifically required during the excystment process. Reverse transcription-PCR experiments showed that both genes display differential expression between the cyst and the vegetative cells. A phylogenetic analysis showed for the first time that the cathepsin B tree is paraphyletic and that the diverging S. histriomuscorum cathepsin B is closely related to its Giardia homologues, which take part in the cyst wall breakdown process. The deduced cathepsin L-like protein sequence displays the structural signatures and phylogenetic relationships of cathepsin H, a protein that is known only in plants and animals and that is involved in the degradation of extracellular matrix components in cancer diseases. The deduced calpain-like protein sequence does not display the calcium-binding domain of conventional calpains; it belongs to a diverging phylogenetic cluster that includes Aspergillus palB, a protein which is involved in a signal transduction pathway that is sensitive to ambient pH.
Sigma-Aldrich
CA-074, ≥98% (HPLC)