- 2,6-Difluorophenol as a bioisostere of a carboxylic acid: bioisosteric analogues of gamma-aminobutyric acid.
2,6-Difluorophenol as a bioisostere of a carboxylic acid: bioisosteric analogues of gamma-aminobutyric acid.
Journal of medicinal chemistry (1999-02-02)
J Qiu, S H Stevenson, M J O'Beirne, R B Silverman
PMID9925739
ABSTRACT
3-(Aminomethyl)-2,6-difluorophenol (6) and 4-(aminomethyl)-2, 6-difluorophenol (7) were synthesized in eight and four steps, respectively, starting from 2,6-difluorophenol, to test the potential of the 2,6-difluorophenol moiety to act as a lipophilic bioisostere of a carboxylic acid. Compounds 6 and 7 are potential bioisosteric analogues of gamma-aminobutyric acid (GABA). Substrate studies and inhibition studies were carried out with pig brain gamma-aminobutyric acid aminotransferase; 6 and 7 are very poor substrates, but both inhibit the enzyme, indicating that the 2, 6-difluorophenol moiety appears to be able to substitute for a carboxylic acid to increase the lipophilicity of drug candidates.