Merck
[Immunochemical therapy of warts and growths due to papillomavirus].
Romanian journal of virology (1993-01-01)
V Topciu, I Fufezan, R Mihăilescu, N Stanciu
ABSTRACT
A complex immuno-chemical treatment (immuno-stimulation, blockage of virus replication, interferon induction) was applied to 23 patients with warts and vegetations with various localisations. Results confirmed the treatment efficiency.
Sigma-Aldrich
Moroxydine hydrochloride, 99%