Merck
  • Determination of mitomycin C in human aqueous humor and serum by high-performance liquid chromatography.
Determination of mitomycin C in human aqueous humor and serum by high-performance liquid chromatography.
Journal of chromatography (1993-09-08)
W Y Li, S K Seah, R T Koda
ABSTRACT
Mitomycin C (MMC) is used in the treatment of disseminated adenocarcinoma of the stomach and pancreas and is used in ophthalmology as adjunctive therapy in trabeculectomy. Since only small volumes of aqueous humor are available for analysis, a sensitive method requiring limited sample preparation was developed. An internal standard, 4-aminoacetophenone, was added to aqueous humor specimens, and the solution was directly injected into the high-performance liquid chromatographic (HPLC) column. The use of a short 50-mm C18 reversed-phase column gave adequate resolution of peaks with improved sensitivity. The method was applicable for determination of MMC in serum, although solid-phase extraction for sample clean-up was required prior to injection into the HPLC column, and analytical columns of 150-250 mm were necessary for adequate resolution of peaks. The method has been validated and is linear from 6.25 to 50 ng/ml in aqueous humor and from 10 to 500 ng/ml in serum.
Sigma-Aldrich
2′-Aminoacetophenone, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
2′-Aminoacetophenone, ≥98%
Pricing and availability is not currently available.