Merck
  • Influence of methylheptenone and related phytoplankton norcarotenoids on heterotrophic aquatic bacteria.
Influence of methylheptenone and related phytoplankton norcarotenoids on heterotrophic aquatic bacteria.
Canadian journal of microbiology (1981-01-01)
W Reichardt
ABSTRACT
Certain norcarotenoids, which have recently been found as excretion products of freshwater cyanobacteria and algae, are potent inhibitors of different metabolic functions in heterotrophic bacteria. 6-Methylhept-5-en-2-one showed the strongest effects and acted as a noncompetitive inhibitor of both glucose uptake and respiration by aquatic isolates of Chromobacterium lividum and Arthrobacter sp. Inhibition of the heterotrophic potential of glucose uptake by 6-methylhept-5-en-2-one was characterized by considerably lower inhibitor constants for bacterial cultures (Ki = 0.4 and 1.6 microM) than for a river water sample (Ki = 186.0 microM).
Sigma-Aldrich
6-Methyl-5-hepten-2-one, natural, ≥98%, FCC
Sigma-Aldrich
6-Methyl-5-hepten-2-one, 99%
Sigma-Aldrich
6-Methyl-5-hepten-2-one, ≥98%, FCC, FG