Merck
  • Primary structure determination of seven novel N-linked carbohydrate chains derived from hemocyanin of Lymnaea stagnalis. 3-O-methyl-D-galactose and N-acetyl-D-galactosamine as constituents of xylose-containing N-linked oligosaccharides in an animal glycoprotein.
Primary structure determination of seven novel N-linked carbohydrate chains derived from hemocyanin of Lymnaea stagnalis. 3-O-methyl-D-galactose and N-acetyl-D-galactosamine as constituents of xylose-containing N-linked oligosaccharides in an animal glycoprotein.
European journal of biochemistry (1987-12-01)
J A Van Kuik, R P Sijbesma, J P Kamerling, J F Vliegenthart, E J Wood
ABSTRACT
Hemocyanin from the freshwater snail Lymnaea stagnalis is a high-molecular-mass copper-containing glyco-protein which functions as oxygen carrier in the hemolymph. To release the carbohydrate chains, the protein was digested by pronase followed by hydrazinolysis and reduction. The oligosaccharide-alditols were purified by gel permeation chromatography on Bio-Gel P-4, followed by HPLC on a Lichrosorb-NH2 column. Using 500-MHz 1H-NMR spectroscopy, in conjunction with sugar, methylation and deamination analysis, the following seven novel primary oligosaccharide structures could be unravelled. (Formula: see text).
Sigma-Aldrich
Methyl α-D-galactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Methyl-β-D-galactopyranoside