Merck
  • Aqueous degradation of N-(hydroxymethyl)phthalimide in the presence of specific and general bases. Kinetic assessment of N-hydroxymethyl derivatives of nitrogen heterocycles as possible prodrugs.
Aqueous degradation of N-(hydroxymethyl)phthalimide in the presence of specific and general bases. Kinetic assessment of N-hydroxymethyl derivatives of nitrogen heterocycles as possible prodrugs.
Journal of pharmaceutical and biomedical analysis (1989-01-01)
M N Khan
ABSTRACT
The conversion of N-(hydroxymethyl)phthalimide (NHPH) to phthalimide could not be detected within 300 s at pH 9.0, whereas in 0.18 M NaOH complete conversion of NHPH to phthalimide was observed within 50 s. In the presence of 0.2-0.4 M 1,4-diazabicyclo[2.2.2]octane buffer solutions (pH 9.30-9.54), 40-60% conversion of NHPH to phthalimide occurred within 90-120 s. The initial concentration of NHPH affected the extent of conversion of NHPH to phthalimide.
Sigma-Aldrich
(S)-(−)-1,2-Diaminopropane dihydrochloride, 99%
Sigma-Aldrich
N-(Hydroxymethyl)phthalimide, 97%
Sigma-Aldrich
(R)-(+)-1,2-Diaminopropane dihydrochloride, 99%
Sigma-Aldrich
1,2-Diaminopropane, 99%