Merck
  • Paroxetine in the treatment of dysthymic disorder without co-morbidities: A double-blind, placebo-controlled, flexible-dose study.
Paroxetine in the treatment of dysthymic disorder without co-morbidities: A double-blind, placebo-controlled, flexible-dose study.
Asian journal of psychiatry (2013-03-08)
Arun V Ravindran, Colin Cameron, Raj Bhatla, Lakshmi N Ravindran, Tricia L da Silva
ABSTRACT
Few published studies have evaluated selective serotonin reuptake inhibitors in dysthymia without current co-morbid major depression. In this 12-week study, 40 dysthymic patients were randomly assigned to either placebo (n=19) or 20-40 mg/day of paroxetine (n=21). At endpoint, the paroxetine group showed significantly greater improvement on the Clinical Global Impression Scale, Beck Depression Inventory, and Quality of Life Enjoyment and Satisfaction Questionnaire (p<0.05), and a trend to superiority over placebo on the Hamilton Depression Rating Scale. Response and remission were significantly higher with paroxetine than placebo (p<0.05). There were no significant differences in drop out rates or frequency of adverse effects, except for excessive sweating (greater with paroxetine, p=0.04). Reporting of multiple side effects was also higher with paroxetine than with placebo (p=0.02). Paroxetine is more effective than placebo in improving symptoms and quality of life in dysthymia, and is generally tolerable.
Sigma-Aldrich
Paroxetine hydrochloride hemihydrate, ≥98% (HPLC), powder
Paroxetine hydrochloride hemihydrate, European Pharmacopoeia (EP) Reference Standard
Paroxetine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Paroxetine maleate salt, ≥98% (HPLC), solid
Supelco
Paroxetine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®